siRNA Details
Gene Symbol: CFC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079530.1 | NP_001072998.1 | 3 | 519 | ||
XM_005263737.4 | XP_005263794.1 | 3 | 401 | |||
GenBank mRNA | BQ777283.1 | - | 3 | 361 | ||
BU784123.1 | - | 3 | 374 |
Gene Symbol: CFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270420.1 | NP_001257349.1 | 3 | 416 | ||
NM_001270421.1 | NP_001257350.1 | 3 | 416 | |||
NM_032545.3 | NP_115934.1 | 3 | 416 | |||
XM_011511486.2 | XP_011509788.1 | 3 | 411 | |||
GenBank mRNA | AF312769.1 | - | 3 | 174 | ||
AK315326.1 | - | 3 | 416 | |||
BC069508.1 | - | 3 | 198 | 7262189 | ||
BC074825.2 | - | 3 | 177 | 30915323 | ||
BC074826.2 | - | 3 | 177 | 30915540 | ||
BC110080.1 | - | 3 | 289 | 40030641 | ||
BC146897.1 | - | 3 | 293 | 9056825 | ||
BM509218.1 | - | 3 | 350 | |||
CA777249.1 | - | 3 | 389 | |||
CA941621.1 | - | 3 | 348 |
