siRNA Details
| Gene Symbol: DGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032564.4 | NP_115953.2 | 2 | 456 | ||
| XM_011545304.2 | XP_011543606.1 | 2 | 160 | |||
| GenBank mRNA | AB048286.2 | - | 2 | 427 | ||
| AF384161.2 | - | 2 | 204 | |||
| AL834287.1 | - | 2 | 335 | |||
| AY358532.1 | - | 2 | 381 | |||
| BC015234.1 | - | 2 | 445 | 3903313 | ||
| DB462004.1 | - | 2 | 456 | |||

DGAT2
Genomic Map