siRNA Details
Gene Symbol: FAM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032648.2 | NP_116037.2 | 2 | 818 | ||
GenBank mRNA | BC004269.1 | - | 2 | 802 | 3613742 | |
BX091465.1 | - | 1 | 50 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032648.2 | NP_116037.2 | 2 | 818 | ||
GenBank mRNA | BC004269.1 | - | 2 | 802 | 3613742 | |
BX091465.1 | - | 1 | 50 |