siRNA Details
| Gene Symbol: FAM71F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282788.1 | NP_001269717.1 | 3 | 573 | ||
| NM_032599.3 | NP_115988.1 | 3 | 573 | |||
| NR_104242.1 | 3 | 573 | ||||
| XM_017012743.1 | XP_016868232.1 | 3 | 616 | |||
| GenBank mRNA | AF367470.1 | - | 3 | 526 | ||
| AK310308.1 | - | 3 | 520 | |||
| AK310426.1 | - | 4 | 429 | |||
| AK313608.1 | - | 3 | 524 | |||
| BC037248.1 | - | 3 | 512 | 5165912 | ||

FAM71F1
Genomic Map