siRNA Details
| Gene Symbol: KBTBD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032505.2 | NP_115894.2 | 3 | 999 | ||
| XM_005264771.3 | XP_005264828.1 | 2 | 784 | |||
| GenBank mRNA | AB058745.1 | - | 1 | 722 | ||
| AF385438.1 | - | 2 | 932 | |||
| AK096640.1 | - | 3 | 995 | |||
| BC117487.1 | - | 2 | 896 | 40126038 | ||

KBTBD8
Genomic Map