siRNA Details
| Gene Symbol: TMEM185A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174092.2 | NP_001167563.1 | 6 | 2405 | ||
| NM_032508.3 | NP_115897.1 | 7 | 2582 | |||
| NR_104121.1 | 4 | 2113 | ||||
| GenBank mRNA | AF353675.1 | - | 4 | 1810 | ||
| AF530473.1 | - | 8 | 2563 | |||
| AK096308.1 | - | 7 | 2396 | |||
| AK097391.1 | - | 4 | 1920 | |||
| AK128688.1 | - | 4 | 3406 | |||
| BC013793.1 | - | 4 | 1656 | |||

TMEM185A
Genomic Map