siRNA Details
| Gene Symbol: C22orf23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207062.1 | NP_001193991.1 | 6 | 987 | ||
| NM_032561.4 | NP_115950.3 | 7 | 1198 | |||
| XM_005261781.1 | XP_005261838.1 | 6 | 1332 | |||
| XM_005261782.3 | XP_005261839.1 | 6 | 1212 | |||
| XM_005261783.2 | XP_005261840.1 | 5 | 1269 | |||
| XM_005261784.2 | XP_005261841.1 | 6 | 1099 | |||
| GenBank mRNA | AF324466.1 | - | 6 | 998 | ||
| AK057349.1 | - | 7 | 1248 | |||
| BC031998.1 | - | 7 | 1080 | 4755328 | ||

C22orf23
Genomic Map