siRNA Details
| Gene Symbol: FAM167B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032648.2 | NP_116037.2 | 2 | 831 | ||
| GenBank mRNA | BC004269.1 | - | 2 | 815 | 3613742 | |
| BX091465.1 | - | 1 | 63 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM167B| Gene Symbol: FAM167B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032648.2 | NP_116037.2 | 2 | 831 | ||
| GenBank mRNA | BC004269.1 | - | 2 | 815 | 3613742 | |
| BX091465.1 | - | 1 | 63 | |||