siRNA Details
| Gene Symbol: INF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031714.3 | NP_001026884.3 | 3 | 577 | ||
| NM_022489.3 | NP_071934.3 | 3 | 577 | |||
| NM_032714.2 | NP_116103.1 | 3 | 577 | |||
| XM_005268004.4 | XP_005268061.1 | 3 | 664 | |||
| XM_005268005.4 | XP_005268062.1 | 3 | 635 | |||
| XM_017021595.1 | XP_016877084.1 | 3 | 635 | |||
| GenBank mRNA | AK290083.1 | - | 3 | 562 | ||
| BC006173.1 | - | 3 | 562 | |||
| BI489810.1 | - | 3 | 577 | |||
| BX248757.1 | - | 3 | 546 | |||

INF2
Genomic Map