siRNA Details
| Gene Symbol: PERM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291366.1 | NP_001278295.1 | 4 | 2731 | ||
| NM_001291367.1 | NP_001278296.1 | 5 | 2378 | |||
| XM_017002583.1 | XP_016858072.1 | 4 | 2833 | |||
| XM_017002584.1 | XP_016858073.1 | 4 | 2856 | |||
| XM_017002585.1 | XP_016858074.1 | 3 | 3222 | |||
| GenBank mRNA | AK123855.1 | - | 5 | 2354 | ||
| BC006300.2 | - | 2 | 528 | |||

PERM1
Genomic Map