siRNA Details
| Gene Symbol: MINA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042533.2 | NP_001035998.1 | 2 | 753 | ||
| NM_001261829.1 | NP_001248758.1 | 2 | 286 | |||
| NM_032778.5 | NP_116167.3 | 2 | 753 | |||
| NM_153182.3 | NP_694822.2 | 2 | 286 | |||
| XM_005247838.4 | XP_005247895.1 | 2 | 503 | |||
| GenBank mRNA | AB083189.1 | - | 2 | 389 | ||
| AB083190.1 | - | 2 | 389 | |||
| AB083191.1 | - | 1 | 206 | |||
| AB083192.1 | - | 2 | 275 | |||
| AB083193.1 | - | 2 | 432 | |||
| AK027299.1 | - | 2 | 236 | |||
| AL550737.3 | - | 2 | 682 | |||
| AY302110.3 | - | 2 | 456 | |||
| AY390536.1 | - | 2 | 438 | |||
| BC014928.1 | - | 2 | 440 | 3916148 | ||
| CR627479.1 | - | 1 | 39 | |||
| DA696177.1 | - | 2 | 286 | |||
| DQ453796.1 | - | 2 | 250 | |||

MINA
Genomic Map