siRNA Details
| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK027370.1 | - | 3 | 281 | ||
| AK128537.1 | - | 3 | 266 | |||
| BC030549.1 | - | 3 | 298 | |||
| BC132961.1 | - | 3 | 275 | 40146983 | ||
| BC136542.1 | - | 3 | 275 | 9020532 | ||
| BC144235.1 | - | 3 | 275 | |||

PARP10
Genomic Map