siRNA Details
| Gene Symbol: CEP89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032816.4 | NP_116205.3 | 8 | 991 | ||
| XM_005259344.3 | XP_005259401.1 | 8 | 986 | |||
| XM_011527425.2 | XP_011525727.1 | 5 | 438 | |||
| XM_017027398.1 | XP_016882887.1 | 8 | 986 | |||
| GenBank mRNA | AK027546.1 | - | 7 | 692 | ||
| AL832158.1 | - | 8 | 918 | |||
| BC032307.1 | - | 8 | 925 | |||
| BC136328.1 | - | 8 | 906 | 9020315 | ||

CEP89
Genomic Map