siRNA Details
| Gene Symbol: USP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301175.1 | NP_001288104.1 | 7 | 899 | ||
| NM_032663.4 | NP_116052.2 | 4 | 525 | |||
| XM_005253962.3 | XP_005254019.1 | 4 | 511 | |||
| XM_005253965.4 | XP_005254022.1 | 3 | 336 | |||
| XM_006719653.3 | XP_006719716.1 | 5 | 598 | |||
| XM_011538894.2 | XP_011537196.1 | 2 | 232 | |||
| XM_017020048.1 | XP_016875537.1 | 4 | 520 | |||
| XM_017020049.1 | XP_016875538.1 | 4 | 511 | |||
| XM_017020050.1 | XP_016875539.1 | 4 | 620 | |||
| XM_017020051.1 | XP_016875540.1 | 5 | 476 | |||
| XM_017020052.1 | XP_016875541.1 | 4 | 522 | |||
| XM_017020053.1 | XP_016875542.1 | 3 | 252 | |||
| XM_017020054.1 | XP_016875543.1 | 2 | 130 | |||
| GenBank mRNA | AJ586136.1 | - | 4 | 416 | ||
| AK027820.1 | - | 4 | 490 | |||
| AK097830.1 | - | 7 | 899 | |||
| BC004868.2 | - | 4 | 515 | 3833242 | ||
| BC022094.2 | - | 2 | 67 | |||
| HY127211.1 | - | 2 | 233 | |||

USP30
Genomic Map