siRNA Details
| Gene Symbol: ABHD14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146314.1 | NP_001139786.1 | 4 | 1814 | ||
| NM_001254753.1 | NP_001241682.1 | 3 | 1575 | |||
| NM_032750.2 | NP_116139.1 | 4 | 2008 | |||
| GenBank mRNA | AK075112.1 | - | 4 | 1399 | ||
| AK092152.1 | - | 1 | 2199 | |||
| AK096073.1 | - | 3 | 1575 | |||
| AK296599.1 | - | 4 | 1069 | |||
| AK298022.1 | - | 5 | 1146 | |||
| BC007234.1 | - | 4 | 1392 | 2823284 | ||
| BC050650.1 | - | 4 | 1206 | |||
| BC056411.1 | - | 4 | 1215 | 6048106 | ||
| BC071931.1 | - | 5 | 1627 | |||

ABHD14B
Genomic Map