siRNA Details
| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026827.1 | 5 | 1097 | |||
| GenBank mRNA | AK024638.1 | - | 1 | 1685 | ||
| AK090789.1 | - | 5 | 1044 | |||
| AK092104.1 | - | 5 | 1047 | |||
| BC007394.2 | - | 4 | 787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00839| Gene Symbol: LINC00839 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026827.1 | 5 | 1097 | |||
| GenBank mRNA | AK024638.1 | - | 1 | 1685 | ||
| AK090789.1 | - | 5 | 1044 | |||
| AK092104.1 | - | 5 | 1047 | |||
| BC007394.2 | - | 4 | 787 | |||