siRNA Details
| Gene Symbol: MIGA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032809.2 | NP_116198.2 | 16 | 2796 | ||
| XM_005252282.1 | XP_005252339.1 | 16 | 2815 | |||
| XM_011519117.1 | XP_011517419.1 | 16 | 2836 | |||
| XM_011519118.1 | XP_011517420.1 | 16 | 2745 | |||
| GenBank mRNA | AJ420465.1 | - | 1 | 239 | ||
| AK027502.1 | - | 8 | 1690 | |||
| AK054974.1 | - | 2 | 1742 | |||
| AK074127.1 | - | 17 | 3558 | |||
| AK075421.1 | - | 14 | 2808 | |||
| BC009114.1 | - | 16 | 2796 | 4153683 | ||

MIGA2
Genomic Map