siRNA Details
| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282736.1 | NP_001269665.1 | 2 | 836 | ||
| NM_032812.8 | NP_116201.7 | 2 | 836 | |||
| XM_011519750.2 | XP_011518052.1 | 2 | 1064 | |||
| GenBank mRNA | AF378757.1 | - | 2 | 208 | ||
| AK304591.1 | - | 3 | 720 | |||
| AY358486.1 | - | 2 | 836 | |||
| BC012885.1 | - | 2 | 442 | 3865073 | ||

PLXDC2
Genomic Map