siRNA Details
| Gene Symbol: RNFT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109903.1 | NP_001103373.1 | 5, 6 | 868 | ||
| NM_032814.3 | NP_116203.2 | 5, 6 | 868 | |||
| GenBank mRNA | AK027533.1 | - | 6, 7 | 904 | ||
| BC011878.2 | - | 5, 6 | 848 | 3532898 | ||
| BQ676121.1 | - | 2, 3 | 539 | |||

RNFT2
Genomic Map