siRNA Details
| Gene Symbol: RTN4IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318746.1 | NP_001305675.1 | 9 | 1064 | ||
| NM_032730.5 | NP_116119.2 | 9 | 1650 | |||
| XM_011536192.2 | XP_011534494.1 | 10 | 1738 | |||
| XM_017011376.1 | XP_016866865.1 | 8 | 1466 | |||
| GenBank mRNA | AK095207.1 | - | 9 | 1064 | ||
| AK313962.1 | - | 9 | 1568 | |||
| BC006399.2 | - | 9 | 1445 | 4309263 | ||

RTN4IP1
Genomic Map