siRNA Details
| Gene Symbol: ADTRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143948.1 | NP_001137420.1 | 7 | 1374 | ||
| NM_032744.3 | NP_116133.1 | 6 | 1320 | |||
| XM_005249454.3 | XP_005249511.1 | 7 | 1345 | |||
| XM_011514956.1 | XP_011513258.1 | 8 | 1451 | |||
| XM_017011377.1 | XP_016866866.1 | 7 | 1044 | |||
| GenBank mRNA | AJ420520.1 | - | 6 | 1089 | ||
| AK123258.1 | - | 5 | 1454 | |||
| AK310611.1 | - | 3 | 756 | |||
| BC007011.1 | - | 6 | 1108 | 3686576 | ||

ADTRP
Genomic Map