siRNA Details
| Gene Symbol: LINC00525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038407.1 | 3 | 889 | |||
| GenBank mRNA | BC007354.2 | - | 3 | 889 | 3616854 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00525| Gene Symbol: LINC00525 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038407.1 | 3 | 889 | |||
| GenBank mRNA | BC007354.2 | - | 3 | 889 | 3616854 | |