siRNA Details
| Gene Symbol: PLXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282736.1 | NP_001269665.1 | 5 | 1203 | ||
| NM_032812.8 | NP_116201.7 | 6 | 1350 | |||
| XM_011519750.2 | XP_011518052.1 | 6 | 1578 | |||
| GenBank mRNA | AF378757.1 | - | 6 | 722 | ||
| AK027529.1 | - | 1 | 63 | |||
| AK304591.1 | - | 7 | 1234 | |||
| AY358486.1 | - | 6 | 1350 | |||
| BC012885.1 | - | 5 | 809 | 3865073 | ||

PLXDC2
Genomic Map