siRNA Details
| Gene Symbol: CEP89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032816.4 | NP_116205.3 | 17 | 2062 | ||
| XM_005259344.3 | XP_005259401.1 | 17 | 1985 | |||
| XM_011527425.2 | XP_011525727.1 | 14 | 1509 | |||
| GenBank mRNA | AK027546.1 | - | 16 | 1763 | ||
| AL832158.1 | - | 16 | 1885 | |||
| BC136328.1 | - | 17 | 1977 | 9020315 | ||
| BE208545.1 | - | 3 | 348 | |||
| DB539713.2 | - | 2 | 83 | |||

CEP89
Genomic Map