siRNA Details
| Gene Symbol: FRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166133.1 | NP_001159605.1 | 11, 12 | 1558 | ||
| NM_025074.6 | NP_079350.5 | 11, 12 | 1558 | |||
| XM_006714316.2 | XP_006714379.1 | 11, 12 | 1805 | |||
| GenBank mRNA | AK027833.1 | - | 6, 7 | 543 | ||
| BC020192.1 | - | 3, 4 | 338 | |||
| BC052281.1 | - | 6, 7 | 645 | |||
| BC064487.1 | - | 6, 7 | 645 | |||
| BC094853.1 | - | 3, 4 | 338 | |||
| BC131820.1 | - | 11, 12 | 1148 | |||

FRAS1
Genomic Map