siRNA Details
| Gene Symbol: PNMA6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032882.5 | NP_116271.3 | 2 | 413 | ||
| GenBank mRNA | AK055322.1 | - | 2 | 345 | ||
| AK289879.1 | - | 2 | 391 | |||
| BC007631.1 | - | 1 | 145 | 3507248 | ||
| BX423799.2 | - | 2 | 360 | |||
| DA091739.1 | - | 2 | 371 | |||
| DB172169.1 | - | 2 | 409 | |||

PNMA6A
Genomic Map