siRNA Details
| Gene Symbol: ATG4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281504.1 | NP_001268433.1 | 5, 6 | 1104 | ||
| NM_032885.5 | NP_116274.3 | 5, 6 | 1030 | |||
| NR_104024.1 | 5, 6 | 986 | ||||
| NR_104025.1 | 4, 5 | 753 | ||||
| GenBank mRNA | AJ312332.1 | - | 5, 6 | 846 | ||
| AK056210.1 | - | 5, 6 | 1005 | |||
| AK223383.1 | - | 5, 6 | 955 | |||
| AK294676.1 | - | 5, 6 | 1104 | |||
| AK314712.1 | - | 5, 6 | 965 | |||
| BC007639.1 | - | 4, 5 | 676 | |||
| BC068992.1 | - | 5, 6 | 1006 | 6578102 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_153583, AK081605, AK081002, AK036571, AK036282, and others.

ATG4D
Genomic Map