siRNA Details
| Gene Symbol: ZNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278284.1 | NP_001265213.1 | 6 | 737 | ||
| NM_001278287.1 | NP_001265216.1 | 6 | 653 | |||
| NM_001278290.1 | NP_001265219.1 | 5 | 498 | |||
| NM_001278291.1 | NP_001265220.1 | 4 | 367 | |||
| NM_001278292.1 | NP_001265221.1 | 5 | 488 | |||
| NM_001318135.1 | NP_001305064.1 | 6 | 615 | |||
| NM_001318136.1 | NP_001305065.1 | 3 | 647 | |||
| NM_032924.4 | NP_116313.3 | 6 | 619 | |||
| XM_006716111.1 | XP_006716174.1 | 6 | 673 | |||
| XM_017012582.1 | XP_016868071.1 | 6 | 687 | |||
| XM_017012583.1 | XP_016868072.1 | 4 | 836 | |||
| GenBank mRNA | AK000223.1 | - | 4 | 354 | ||
| AK091984.1 | - | 3 | 619 | |||
| AK315479.1 | - | 4 | 333 | |||
| BC011887.2 | - | 6 | 543 | 3831842 | ||
| BC013603.1 | - | 6 | 562 | 3865426 | ||
| BC025265.1 | - | 6 | 552 | |||
| BQ071498.1 | - | 6 | 449 | |||
| BT006918.1 | - | 3 | 184 | |||
| BX647182.1 | - | 5 | 1260 | |||
| CB988353.1 | - | 3 | 647 | |||
| DA443888.1 | - | 4 | 353 | |||
| DA623743.1 | - | 5 | 462 | |||
| X60153.1 | - | 4 | 292 | |||

ZNF3
Genomic Map