siRNA Details
| Gene Symbol: ARRDC1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122035.1 | 3 | 769 | |||
| NR_122036.1 | 3 | 458 | ||||
| GenBank mRNA | AF267857.1 | - | 3 | 687 | ||
| BC011781.2 | - | 3 | 671 | |||
| BC066646.1 | - | 3 | 476 | 5749061 | ||
| BC117267.1 | - | 2 | 229 | 40125818 | ||
| CX164735.1 | - | 3 | 458 | |||

ARRDC1-AS1
Genomic Map