siRNA Details
| Gene Symbol: NXF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032946.2 | NP_116564.2 | 15 | 1297 | ||
| NR_028089.1 | 15 | 1297 | ||||
| GenBank mRNA | AJ277654.1 | - | 15 | 1297 | ||
| AJ277655.1 | - | 15 | 1297 | |||
| AJ277656.1 | - | 15 | 1233 | |||
| AJ277657.1 | - | 13 | 1108 | |||
| AJ277658.1 | - | 13 | 1108 | |||
| BC131708.1 | - | 14 | 1118 | 40121743 | ||

NXF5
Genomic Map