siRNA Details
| Gene Symbol: FIZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032836.2 | NP_116225.2 | 3 | 2040 | ||
| XM_005259352.4 | XP_005259409.1 | 3 | 3065 | |||
| XM_011527426.2 | XP_011525728.1 | 3 | 2001 | |||
| GenBank mRNA | AK027674.1 | - | 3 | 2040 | ||
| BC132777.1 | - | 2 | 2001 | 40146799 | ||
| BC136512.1 | - | 2 | 1999 | 9020502 | ||

FIZ1
Genomic Map