siRNA Details
| Gene Symbol: SPRYD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032840.2 | NP_116229.1 | 2 | 229 | ||
| GenBank mRNA | AK027706.1 | - | 2 | 163 | ||
| AK074694.1 | - | 2 | 195 | |||
| AK294092.1 | - | 2 | 213 | |||
| BC136312.1 | - | 2 | 162 | 9020299 | ||
| DA389433.1 | - | 2 | 229 | |||

SPRYD3
Genomic Map