siRNA Details
| Gene Symbol: CCDC183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039374.4 | NP_001034463.4 | 6, 7 | 736 | ||
| GenBank mRNA | AK090683.1 | - | 2, 3 | 312 | ||
| AK092639.1 | - | 3, 4 | 994 | |||
| BC137317.1 | - | 6, 7 | 711 | 9021314 | ||
| BC137318.1 | - | 6, 7 | 711 | 9021315 | ||
| BF979345.1 | - | 652 | ||||
| DA235138.1 | - | 2, 3 | 266 | |||

CCDC183
Genomic Map