siRNA Details
Gene Symbol: TMEM141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032928.3 | NP_116317.1 | 4, 5 | 359 | ||
GenBank mRNA | BC007834.2 | - | 4, 5 | 329 | 4100086 | |
CB107060.1 | - | 4, 5 | 359 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMEM141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032928.3 | NP_116317.1 | 4, 5 | 359 | ||
GenBank mRNA | BC007834.2 | - | 4, 5 | 329 | 4100086 | |
CB107060.1 | - | 4, 5 | 359 |