siRNA Details
| Gene Symbol: TMEM241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318834.1 | NP_001305763.1 | 16 | 1371 | ||
| NM_032933.5 | NP_116322.3 | 15 | 1256 | |||
| NR_134875.1 | 14 | 1207 | ||||
| NR_134876.1 | 11 | 1127 | ||||
| NR_134877.1 | 14 | 1210 | ||||
| XM_011526233.2 | XP_011524535.1 | 16 | 1348 | |||
| XM_017026045.1 | XP_016881534.1 | 14 | 1170 | |||
| XM_017026049.1 | XP_016881538.1 | 15 | 1252 | |||
| XM_017026050.1 | XP_016881539.1 | 15 | 1274 | |||
| XM_017026051.1 | XP_016881540.1 | 15 | 1176 | |||
| GenBank mRNA | AK123623.1 | - | 11 | 1117 | ||
| AK126247.1 | - | 14 | 1199 | |||
| BC006280.2 | - | 3 | 476 | |||
| BC054507.1 | - | 9 | 753 | |||
| BC082984.1 | - | 3 | 604 | |||
| BC114510.1 | - | 14 | 1143 | 40023922 | ||
| BC114561.1 | - | 15 | 1192 | 40023923 | ||

TMEM241
Genomic Map