siRNA Details
| Gene Symbol: NXF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028089.1 | 16 | 1481 | |||
| GenBank mRNA | AJ277654.1 | - | 16 | 1481 | ||
| AJ277655.1 | - | 16 | 1456 | |||
| AJ277656.1 | - | 16 | 1392 | |||
| AJ277657.1 | - | 14 | 1267 | |||
| AJ277658.1 | - | 14 | 1386 | |||

NXF5
Genomic Map