siRNA Details
| Gene Symbol: BBS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252678.1 | NP_001239607.1 | 3 | 194 | ||
| NM_001320665.1 | NP_001307594.1 | 4 | 246 | |||
| NM_033028.4 | NP_149017.2 | 4 | 246 | |||
| NR_045565.1 | 5 | 304 | ||||
| NR_045566.1 | 4 | 559 | ||||
| XM_011521848.1 | XP_011520150.1 | 4 | 216 | |||
| XM_011521849.1 | XP_011520151.1 | 4 | 221 | |||
| XM_011521851.1 | XP_011520153.1 | 4 | 216 | |||
| XM_017022450.1 | XP_016877939.1 | 4 | 527 | |||
| XM_017022451.1 | XP_016877940.1 | 3 | 187 | |||
| XM_017022452.1 | XP_016877941.1 | 4 | 529 | |||
| XM_017022453.1 | XP_016877942.1 | 4 | 211 | |||
| XM_017022454.1 | XP_016877943.1 | 3 | 161 | |||
| GenBank mRNA | AF359281.1 | - | 4 | 218 | ||
| AK075321.1 | - | 4 | 210 | |||
| AK298922.1 | - | 5 | 276 | |||
| AK302224.1 | - | 4 | 513 | |||
| AK303706.1 | - | 3 | 189 | |||
| AK314507.1 | - | 4 | 218 | |||
| AY457143.1 | - | 4 | 318 | |||
| BC008923.2 | - | 1 | 23 | |||
| BC027624.2 | - | 4 | 216 | 4824276 | ||
| BE727117.1 | - | 3 | 187 | |||
| BX647855.1 | - | 3 | 165 | |||
| DB269838.1 | - | 4 | 239 | |||
| DC320069.1 | - | 4 | 241 | |||
| DC372967.1 | - | 4 | 246 | |||
| KU177936.1 | - | 4 | 197 | |||

BBS4
Genomic Map