siRNA Details
| Gene Symbol: MEDAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032849.3 | NP_116238.2 | 4, 5 | 1136 | ||
| XM_017020801.1 | XP_016876290.1 | 3, 4 | 1516 | |||
| GenBank mRNA | AB055407.1 | - | 3, 4 | 522 | ||
| AK055635.1 | - | 4, 5 | 1120 | |||
| BC040165.1 | - | 4, 5 | 1020 | 5590156 | ||
| BC101624.1 | - | 4, 5 | 874 | 8069130 | ||
| BC101626.1 | - | 4, 5 | 874 | 8069132 | ||

MEDAG
Genomic Map