siRNA Details
| Gene Symbol: SERAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032861.3 | NP_116250.3 | 17 | 2796 | ||
| XM_006715586.2 | XP_006715649.1 | 16 | 2725 | |||
| XM_011536196.2 | XP_011534498.1 | 16 | 2819 | |||
| XM_011536198.2 | XP_011534500.1 | 15 | 2721 | |||
| GenBank mRNA | AK023656.1 | - | 1 | 832 | ||
| AK027823.1 | - | 17 | 2786 | |||
| AK125878.1 | - | 18 | 3988 | |||
| BC001705.1 | - | 17 | 2753 | 3350731 | ||
| BX648104.1 | - | 15 | 4172 | |||

SERAC1
Genomic Map