siRNA Details
| Gene Symbol: CGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252335.1 | NP_001239264.1 | 20 | 4460 | ||
| NM_032866.4 | NP_116255.2 | 19 | 4345 | |||
| XM_005254726.3 | XP_005254783.1 | 19 | 4366 | |||
| XM_005254727.4 | XP_005254784.1 | 20 | 4744 | |||
| XM_011522120.1 | XP_011520422.1 | 20 | 4402 | |||
| XM_011522121.1 | XP_011520423.1 | 19 | 4525 | |||
| XM_017022685.1 | XP_016878174.1 | 19 | 4983 | |||
| XM_017022686.1 | XP_016878175.1 | 19 | 4978 | |||
| GenBank mRNA | AB051536.2 | - | 18 | 3504 | ||
| AK027863.1 | - | 14 | 2255 | |||
| AY274808.1 | - | 19 | 4343 | |||
| BC118918.1 | - | 18 | 4286 | 40074275 | ||

CGNL1
Genomic Map