siRNA Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032898.4 | NP_116287.2 | 3 | 1070 | ||
| XM_005269370.4 | XP_005269427.1 | 3 | 1045 | |||
| XM_011513246.2 | XP_011511548.1 | 3 | 806 | |||
| GenBank mRNA | AK058155.1 | - | 3 | 968 | ||
| BC007827.2 | - | 3 | 999 | 3533603 | ||

CEP19
Genomic Map