siRNA Details
| Gene Symbol: ZNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007147.3 | NP_009078.1 | 4 | 691 | ||
| XM_017027249.1 | XP_016882738.1 | 4 | 709 | |||
| GenBank mRNA | AK292687.1 | - | 4 | 624 | ||
| BC052997.1 | - | 4 | 623 | 6173867 | ||
| D50419.1 | - | 4 | 617 | |||

ZNF175
Genomic Map