siRNA Details
| Gene Symbol: TRIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB362928.1 | - | 1, 2 | 416 | ||
| AF220027.1 | - | 1, 2 | 478 | |||
| AF220028.1 | - | 1, 2 | 478 | |||
| AK027593.1 | - | 2, 3 | 673 | |||
| DQ288685.1 | - | 1, 2 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM5| Gene Symbol: TRIM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB362928.1 | - | 1, 2 | 416 | ||
| AF220027.1 | - | 1, 2 | 478 | |||
| AF220028.1 | - | 1, 2 | 478 | |||
| AK027593.1 | - | 2, 3 | 673 | |||
| DQ288685.1 | - | 1, 2 | 416 | |||