siRNA Details
| Gene Symbol: KRTAP4-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033059.3 | NP_149048.2 | 1 | 743 | ||
| GenBank mRNA | AJ406944.1 | - | 1 | 749 | ||
| BC126131.1 | - | 1 | 725 | 8991847 | ||
| BC130562.1 | - | 1 | 725 | 40146593 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-11| Gene Symbol: KRTAP4-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033059.3 | NP_149048.2 | 1 | 743 | ||
| GenBank mRNA | AJ406944.1 | - | 1 | 749 | ||
| BC126131.1 | - | 1 | 725 | 8991847 | ||
| BC130562.1 | - | 1 | 725 | 40146593 | ||