siRNA Details
| Gene Symbol: KRTAP4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033060.2 | NP_149049.1 | 2 | 567 | ||
| GenBank mRNA | AJ406942.1 | - | 2 | 567 | ||
| BC065372.1 | - | 3 | 1064 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-1| Gene Symbol: KRTAP4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033060.2 | NP_149049.1 | 2 | 567 | ||
| GenBank mRNA | AJ406942.1 | - | 2 | 567 | ||
| BC065372.1 | - | 3 | 1064 | |||