siRNA Details
| Gene Symbol: KRTAP4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033062.3 | NP_149051.1 | 1 | 539 | ||
| GenBank mRNA | AJ406934.1 | - | 1 | 539 | ||
| BC127252.1 | - | 1 | 519 | 40030890 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP4-2| Gene Symbol: KRTAP4-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033062.3 | NP_149051.1 | 1 | 539 | ||
| GenBank mRNA | AJ406934.1 | - | 1 | 539 | ||
| BC127252.1 | - | 1 | 519 | 40030890 | ||