siRNA Details
| Gene Symbol: ZCRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033114.3 | NP_149105.3 | 3, 4 | 331 | ||
| XM_017020124.1 | XP_016875613.1 | 2, 3 | 222 | |||
| GenBank mRNA | AB062361.1 | - | 3, 4 | 331 | ||
| AK308244.1 | - | 3, 4 | 201 | |||
| BC005099.1 | - | 3, 4 | 281 | |||
| BC010177.2 | - | 3, 4 | 296 | |||
| BC022543.1 | - | 3, 4 | 306 | 4797962 | ||

ZCRB1
Genomic Map