siRNA Details
| Gene Symbol: LACRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033277.1 | NP_150593.1 | 4 | 353 | ||
| GenBank mRNA | AF238867.1 | - | 4 | 367 | ||
| BC062217.1 | - | 4 | 373 | 30327037 | ||
| BC069317.1 | - | 4 | 353 | 7262178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LACRT| Gene Symbol: LACRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033277.1 | NP_150593.1 | 4 | 353 | ||
| GenBank mRNA | AF238867.1 | - | 4 | 367 | ||
| BC062217.1 | - | 4 | 373 | 30327037 | ||
| BC069317.1 | - | 4 | 353 | 7262178 | ||