siRNA Details
| Gene Symbol: FANCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018115.2 | NP_001018125.1 | 3 | 294 | ||
| NM_001319984.1 | NP_001306913.1 | 3 | 366 | |||
| NM_033084.4 | NP_149075.2 | 3 | 294 | |||
| GenBank mRNA | AF230336.1 | - | 3 | 237 | ||
| AF340183.1 | - | 3 | 237 | |||
| AK307512.1 | - | 4 | 267 | |||
| BC013582.2 | - | 3 | 225 | |||
| BX956311.1 | - | 3 | 265 | |||
| DA749163.1 | - | 4 | 491 | |||
| DA864640.1 | - | 3 | 295 | |||
| DA887207.1 | - | 3 | 290 | |||

FANCD2
Genomic Map