siRNA Details
| Gene Symbol: RRP36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033112.2 | NP_149103.1 | 4 | 412 | ||
| GenBank mRNA | AF161371.1 | - | 3 | 168 | ||
| AK313816.1 | - | 4 | 422 | |||
| BC006293.2 | - | 4 | 406 | |||
| BC011933.2 | - | 4 | 412 | 4300313 | ||

RRP36
Genomic Map